2-Chloro-3-methoxyphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (2-Chloro-3-methoxyphenyl)boronic acid

29289-25G 29289-5G
76901-188EA 548.49 USD
76901-188 76901-190
2-Chloro-3-methoxyphenylboronic acid ≥95%
2-Chloro-3-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BClO₃
MW: 186.4006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12913933
CAS Number: 854778-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR