2-Bromo-3,6-difluorophenol ≥97%

Supplier: AOBChem USA

Warning

29250-1G 29250-25G 29250-5G
76901-176EA 84.88 USD
76901-176 76901-178 76901-180
2-Bromo-3,6-difluorophenol ≥97%
2-Bromo-3,6-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrF₂O
MW: 208.988 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13176518
CAS Number: 1208077-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR