2'-Methoxy-2,2,2-trifluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

29159-25G 29159-5G
76901-148EA 453.01 USD
76901-148 76901-150
2'-Methoxy-2,2,2-trifluoroacetophenone ≥97%
2'-Methoxy-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00461894
CAS Number: 26944-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR