2-Bromo-3,6-difluorotoluene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-1,4-difluoro-3-methylbenzene

29120-1G 29120-25G 29120-500MG 29120-5G
76901-136EA 195.21 USD
76901-136 76901-138 76901-140 76901-142
2-Bromo-3,6-difluorotoluene ≥97%
2-Bromo-3,6-difluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MW: 207.015 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12922624
CAS Number: 1208074-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR