2-Bromo-6-fluoro-4-(trifluoromethyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

29023-1G 29023-25G 29023-500MG 29023-5G
76901-098EA 349.04 USD
76901-098 76901-100 76901-102 76901-104
2-Bromo-6-fluoro-4-(trifluoromethyl)benzaldehyde ≥95%
2-Bromo-6-fluoro-4-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₃BrF₄O
MW: 271.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31665800
CAS Number: 2092468-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR