2,6-Dichloro-3-methylphenol ≥97%

Supplier: AOBChem USA

Warning

28888-1G 28888-25G 28888-5G
76901-078EA 66.84 USD
76901-078 76901-080 76901-082
2,6-Dichloro-3-methylphenol ≥97%
2,6-Dichloro-3-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆Cl₂O
MW: 177.028 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061909
CAS Number: 13481-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR