2-Fluoro-3-(methoxymethoxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

28756-1G 28756-25G 28756-500MG 28756-5G
76901-062EA 194.15 USD
76901-062 76901-064 76901-066 76901-068
2-Fluoro-3-(methoxymethoxy)benzaldehyde ≥95%
2-Fluoro-3-(methoxymethoxy)benzaldehyde
Formula: C₉H₉FO₃
MW: 184.164 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31619422
CAS Number: 2179038-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR