2-Fluoro-5-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Fluoro-5-methoxyphenyl)methanol

28696-25G 28696-5G
76901-036EA 418 USD
76901-036 76901-038
2-Fluoro-5-methoxybenzyl alcohol ≥97%
2-Fluoro-5-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉FO₂
MW: 156.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116337
CAS Number: 161643-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR