2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

28556-1G 28556-25G 28556-500MG 28556-5G
76901-014EA 409.51 USD
76901-014 76901-016 76901-018 76901-020
2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid ≥95%
2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Formula: C₁₅H₂₁BO₄
MW: 276.136 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23379533
CAS Number: 890839-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR