2,3-Difluoro-6-methoxytoluene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methyl-3,4-difluoroanisole

28488-1G 28488-25G 28488-5G 28488-500MG
76901-004EA 94.43 USD
76901-004 76901-006 76901-008 77649-792
2,3-Difluoro-6-methoxytoluene ≥97%
2,3-Difluoro-6-methoxytoluene
Formula: C₈H₈F₂O
MW: 158.145 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12922652
CAS Number: 1208078-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR