(2,4-Dichloropyridin-3-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,4-Dichloropyridine-3-methanol

28429-1G 28429-25G 28429-500MG 28429-5G
76900-994EA 171.87 USD
76900-994 76900-996 76900-998 76901-000
(2,4-Dichloropyridin-3-yl)methanol ≥97%
(2,4-Dichloropyridin-3-yl)methanol
Formula: C₆H₅Cl₂NO
MW: 178.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11847307
CAS Number: 945543-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR