2,4-Dichloropyridine-3-boronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

28402-1G 28402-25G 28402-5G
76900-978EA 73.21 USD
76900-978 76900-980 76900-982
2,4-Dichloropyridine-3-boronic acid pinacol ester ≥97% research grade
2,4-Dichloropyridine-3-boronic acid pinacol ester
Formula: C₁₁H₁₄BCl₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD15144793
CAS Number: 1257651-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR