2-Bromo-5-butylThiophene ≥97%

Supplier: AOBChem USA

Warning

28346-1G 28346-25G 28346-500MG 28346-5G
76900-958EA 131.56 USD
76900-958 76900-960 76900-962 76900-964
2-Bromo-5-butylThiophene ≥97%
2-Bromo-5-butylThiophene
Formula: C₈H₁₁BrS
MW: 219.142 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20486050
CAS Number: 128619-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR