2-Ethyl-6-methylphenol ≥97%

Supplier: AOBChem USA

Warning

28329-25G 28329-5G
76900-946EA 230.22 USD
76900-946 76900-948
2-Ethyl-6-methylphenol ≥97%
2-Ethyl-6-methylphenol

Precaución: for Research Use Only

Formula: C₉H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD00962292
CAS Number: 1687-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR