2-Bromo-4-fluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

28244-25G 28244-5G
76900-932EA 516.66 USD
76900-932 76900-934
2-Bromo-4-fluorobenzamide ≥97%
2-Bromo-4-fluorobenzamide
Formula: C₇H₅BrFNO
Storage Temperature: Ambient
MDL Number: MFCD06409122
CAS Number: 1006-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR