2-Bromo-1-(2,2-diethoxyethoxy)-4-fluorobenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
28225-1G
28225-25G
28225-5G
76900-920EA
96.55
USD
76900-920
76900-922
76900-924
2-Bromo-1-(2,2-diethoxyethoxy)-4-fluorobenzene ≥97%
2-Bromo-1-(2,2-diethoxyethoxy)-4-fluorobenzene
Precaución: for Research Use Only
Formula:
C₁₂H₁₆BrFO₃ MW: 307.156 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD14584680 CAS Number: 253429-18-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...