2,6-Dichloro-4-(trifluoromethyl)phenylboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

27981-1G 27981-25G 27981-500MG 27981-5G
76900-872EA 421.18 USD
76900-872 76900-874 76900-876 76900-878
2,6-Dichloro-4-(trifluoromethyl)phenylboronic acid pinacol ester ≥97% research grade
2,6-Dichloro-4-(trifluoromethyl)phenylboronic acid pinacol ester
Formula: C₁₃H₁₄BCl₂F₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD32631800
CAS Number: 2311855-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR