4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde, 2-Formylthiophene-4-boronic acid pinacol ester

27854-25G 27854-5G
76900-830EA 375.56 USD
76900-830 76900-832
4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde ≥97%
4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₅BO₃S
MW: 238.111 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12756464
CAS Number: 881381-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR