2-Chloro-3,5-difluorophenol ≥97%

Supplier: AOBChem USA

Warning

27796-1G 27796-25G 27796-5G
76900-816EA 79.57 USD
76900-816 76900-818 76900-820
2-Chloro-3,5-difluorophenol ≥97%
2-Chloro-3,5-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₃ClF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00075400
CAS Number: 206986-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR