(2-Bromo-4-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-4-methylbenzyl alcohol

27641-1G 27641-25G 27641-500MG 27641-5G
76900-748EA 201.58 USD
76900-748 76900-750 76900-752 76900-754
(2-Bromo-4-methylphenyl)methanol ≥97%
(2-Bromo-4-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₉BrO
Storage Temperature: Refrigerator
MDL Number: MFCD06202838
CAS Number: 824-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR