2-Chloro-3-methoxyisonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

27588-1G 27588-25G 27588-500MG 27588-5G
76900-736EA 292.81 USD
76900-736 76900-738 76900-740 76900-742
2-Chloro-3-methoxyisonicotinic acid ≥97%
2-Chloro-3-methoxyisonicotinic acid
Formula: C₇H₆ClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD13185828
CAS Number: 1214344-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR