2-Chloro-4-(dimethylamino)pyridine-3-carbaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

27547-1G 27547-500MG 27547-5G 27547-25G
76900-710EA 640.79 USD
76900-710 76900-712 76900-714 77658-394
2-Chloro-4-(dimethylamino)pyridine-3-carbaldehyde ≥97% research grade
2-Chloro-4-(dimethylamino)pyridine-3-carbaldehyde
Formula: C₈H₉ClN₂O
MW: 184.623 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21362342
CAS Number: 1282606-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR