2-Fluoro-3-formylbenzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-3-formylphenylboronic acid, pinacol ester, 2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

27369-1G 27369-25G 27369-5G
76900-678EA 62.6 USD
76900-678 76900-680 76900-682
2-Fluoro-3-formylbenzeneboronic acid pinacol ester ≥97%
2-Fluoro-3-formylbenzeneboronic acid pinacol ester
Formula: C₁₃H₁₆BFO₃
MW: 250.076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17015836
CAS Number: 1112209-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR