2-Methoxymethoxy-4-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

27325-1G 27325-25G 27325-500MG 27325-5G
76900-664EA 374.5 USD
76900-664 76900-666 76900-668 76900-670
2-Methoxymethoxy-4-methylphenylboronic acid ≥97%
2-Methoxymethoxy-4-methylphenylboronic acid
Formula: C₉H₁₃BO₄
MW: 196.008 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31806001
CAS Number: 1236304-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR