2-Bromo-5-carboxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

27291-1G 27291-25G 27291-500MG 27291-5G
76900-646EA 290.69 USD
76900-646 76900-648 76900-650 76900-652
2-Bromo-5-carboxyphenylboronic acid ≥97%
2-Bromo-5-carboxyphenylboronic acid
Formula: C₇H₆BBrO₄
MW: 244.835 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28015446
CAS Number: 1448312-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR