2-Methyl-6-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

27189-1G 27189-25G 27189-5G
76900-612EA 62.6 USD
76900-612 76900-614 76900-616
2-Methyl-6-phenylpyridine ≥97%
2-Methyl-6-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₁N
Storage Temperature: Refrigerator
MDL Number: MFCD00796537
CAS Number: 46181-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR