Dimethyl(5-trifluoromethylpyridin-2-yl)amine ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Dimethylamino-5-(trifluoromethyl)pyridine

27136-1G 27136-25G 27136-5G
76900-602EA 109.28 USD
76900-602 76900-604 76900-606
Dimethyl(5-trifluoromethylpyridin-2-yl)amine ≥97% research grade
Dimethyl(5-trifluoromethylpyridin-2-yl)amine
Formula: C₈H₉F₃N₂
MW: 190.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00766812
CAS Number: 136539-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR