2-Bromo-3-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

27061-1G 27061-25G 27061-500MG 27061-5G
76900-584EA 231.28 USD
76900-584 76900-586 76900-588 76900-590
2-Bromo-3-methoxybenzyl alcohol ≥97%
2-Bromo-3-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18391884
CAS Number: 199436-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR