2,5-Dibromo-3-methoxybenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

27019-1G 27019-25G 27019-5G
76900-562EA 82.76 USD
76900-562 76900-564 76900-566
2,5-Dibromo-3-methoxybenzaldehyde ≥97% research grade
2,5-Dibromo-3-methoxybenzaldehyde
Formula: C₈H₆Br₂O₂
MW: 293.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27756695
CAS Number: 856767-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR