2-Methyl-5-nitrobenzimidazole ≥97%

Supplier: AOBChem USA

Warning

26821-25G 26821-5G
76900-498EA 240.83 USD
76900-498 76900-500
2-Methyl-5-nitrobenzimidazole ≥97%
2-Methyl-5-nitrobenzimidazole

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005599
CAS Number: 1792-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR