3-Amino-2-methoxypyridine-5-boronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine, 3-Amino-2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Methoxy-3-aminopyridine-5-boronic acid pinacol ester, 2-(5-Amino-6-methoxy-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Amino-2-methoxypyridine-5-boronic acid Pinacol ester

26783-25G 26783-5G
76900-486EA 219.61 USD
76900-486 76900-488
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester ≥97% research grade
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester
Formula: C₁₂H₁₉BN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD12923427
CAS Number: 893440-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR