2,4-Difluoro-5-bromophenylboronic acid piancol ester ≥97%

Supplier: AOBChem USA

Warning

26779-1G 26779-25G 26779-5G
76900-476EA 62.6 USD
76900-476 76900-478 76900-480
2,4-Difluoro-5-bromophenylboronic acid piancol ester ≥97%
2,4-Difluoro-5-bromophenylboronic acid piancol ester
Formula: C₁₂H₁₄BBrF₂O₂
MW: 318.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24039984
CAS Number: 1419602-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR