2-Iodo-4-isopropoxy-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

26772-1G 26772-500MG 26772-5G
76900-462EA 498.63 USD
76900-462 76900-464 76900-466
2-Iodo-4-isopropoxy-1-methylbenzene ≥97%
2-Iodo-4-isopropoxy-1-methylbenzene
Formula: C₁₀H₁₃IO
MW: 276.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28142585
CAS Number: 1369834-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR