2-Chloro-4-iodo-6-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

26706-1G 26706-25G 26706-500MG 26706-5G
76900-426EA 215.37 USD
76900-426 76900-428 76900-430 76900-432
2-Chloro-4-iodo-6-methoxypyridine ≥97%
2-Chloro-4-iodo-6-methoxypyridine
Formula: C₆H₅ClINO
MW: 269.467 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16610525
CAS Number: 1227602-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR