2-Amino-3-bromo-5-chlorobenzonitrile ≥95%

Supplier: AOBChem USA

Warning

26504-25G 26504-5G
76900-386EA 524.09 USD
76900-386 76900-388
2-Amino-3-bromo-5-chlorobenzonitrile ≥95%
2-Amino-3-bromo-5-chlorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD08436080
CAS Number: 914636-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR