(2-Bromo-5-methylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-5-methylphenylboronic acid

26369-1G 26369-25G 26369-5G
76900-354EA 82.76 USD
76900-354 76900-356 76900-358
(2-Bromo-5-methylphenyl)boronic acid ≥97% research grade
(2-Bromo-5-methylphenyl)boronic acid
Formula: C₇H₈BBrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD26045274
CAS Number: 1450711-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR