2,5-Difluoro-4-methoxyphenol ≥97% research grade

Supplier: AOBChem USA

Warning

26241-1G 26241-25G 26241-500MG 26241-5G
76900-322EA 320.4 USD
76900-322 76900-324 76900-326 76900-328
2,5-Difluoro-4-methoxyphenol ≥97% research grade
2,5-Difluoro-4-methoxyphenol
Formula: C₇H₆F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD06246893
CAS Number: 1024068-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR