6-Methoxy-m-cresol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Hydroxy-4-methylanisol, 3-Hydroxy-4-methoxytoluene, 2-Methoxy-5-methylphenol, 1-Hydroxy-2-methoxy-5-methylbenzene

26174-25G 26174-5G
76900-266EA 244.01 USD
76900-266 76900-268
6-Methoxy-m-cresol ≥97%
6-Methoxy-m-cresol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040901
CAS Number: 1195-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR