2-Chloro-N-methoxy-N-methylpyridine-3-carboxamide ≥97% research grade

Supplier: AOBChem USA

Warning

26145-25G 26145-5G
76900-258EA 336.31 USD
76900-258 76900-260
2-Chloro-N-methoxy-N-methylpyridine-3-carboxamide ≥97% research grade
2-Chloro-N-methoxy-N-methylpyridine-3-carboxamide
Formula: C₈H₉ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11847245
CAS Number: 488149-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR