2-Fluoro-4-iodopyridin-3-ol 95% research grade

Supplier: AOBChem USA

Warning

26137-1G 26137-500MG 26137-5G 26137-25G
76900-246EA 795.68 USD
76900-246 76900-248 76900-250 77654-524
2-Fluoro-4-iodopyridin-3-ol 95% research grade
2-Fluoro-4-iodopyridin-3-ol
Formula: C₅H₃FINO
Storage Temperature: Refrigerator
MDL Number: MFCD16611041
CAS Number: 1227593-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR