2-Methoxy-5-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

26082-25G 26082-5G
76900-220EA 251.44 USD
76900-220 76900-222
2-Methoxy-5-(trifluoromethyl)benzyl alcohol ≥97%
2-Methoxy-5-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD06203094
CAS Number: 685126-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR