2,6-Dichloro-N,N-dimethylpyridine-4-carboxamide ≥97%

Supplier: AOBChem USA

Warning

25999-1G 25999-25G 25999-500MG 25999-5G
76900-188EA 252.5 USD
76900-188 76900-190 76900-192 76900-194
2,6-Dichloro-N,N-dimethylpyridine-4-carboxamide ≥97%
2,6-Dichloro-N,N-dimethylpyridine-4-carboxamide
Formula: C₈H₈Cl₂N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD11726124
CAS Number: 20373-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR