2,6-Dichloro-4-(trifluoromethoxy)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

25937-1G 25937-25G 25937-500MG 25937-5G
76900-172EA 173.99 USD
76900-172 76900-174 76900-176 76900-178
2,6-Dichloro-4-(trifluoromethoxy)benzyl alcohol ≥97%
2,6-Dichloro-4-(trifluoromethoxy)benzyl alcohol
Formula: C₈H₅Cl₂F₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD06660301
CAS Number: 886503-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR