2-Bromo-4-methylbiphenyl ≥97%

Supplier: AOBChem USA

Warning

25806-1G 25806-25G 25806-500MG 25806-5G
76900-130EA 237.65 USD
76900-130 76900-132 76900-134 76900-136
2-Bromo-4-methylbiphenyl ≥97%
2-Bromo-4-methylbiphenyl
Formula: C₁₃H₁₁Br
MW: 247.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23713149
CAS Number: 29180-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR