2-Isoproxy-3-methylpyridine-5-boronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

25757-1G 25757-25G 25757-500MG 25757-5G
76900-108EA 142.17 USD
76900-108 76900-110 76900-112 76900-114
2-Isoproxy-3-methylpyridine-5-boronic acid pinacol ester ≥97% research grade
2-Isoproxy-3-methylpyridine-5-boronic acid pinacol ester
Formula: C₁₅H₂₄BNO₃
Storage Temperature: Ambient
MDL Number: MFCD16996660
CAS Number: 1375303-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR