2-Amino-5-bromo-3-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-3-methoxypyridin-2-amine

25729-25G 25729-5G
76900-086EA 240.83 USD
76900-086 76900-088
2-Amino-5-bromo-3-methoxypyridine ≥97%
2-Amino-5-bromo-3-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD09909849
CAS Number: 42409-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR