Cyclopentanone-p-toluenesulfonylhydrazone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Cyclopentylidene-1-tosylhydrazine, Cyclopentanone Tosylhydrazone

25690-25G 25690-5G
76900-060EA 346.92 USD
76900-060 76900-062
Cyclopentanone-p-toluenesulfonylhydrazone ≥97%
Cyclopentanone-p-toluenesulfonylhydrazone
Formula: C₁₂H₁₆N₂O₂S
MW: 252.33 g/mol
Storage Temperature: Ambient
CAS Number: 17529-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR