2-Fluoro-(4-methoxycarbonyl)phenylboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: Methyl 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 2-Fluoro-4-(methoxycarbonyl)phenylboronic acid,pinacol ester, 2-Fluoro-4-(methoxycarbonyl)benzeneboronic acid pinacol ester

25554-25G 25554-5G
76900-020EA 289.63 USD
76900-020 76900-022
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid pinacol ester ≥97% research grade
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid pinacol ester
Formula: C₁₄H₁₈BFO₄
MW: 280.0997 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11855962
CAS Number: 603122-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR