2,6-Difluoro-4-(methylthio)benzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

25500-1G 25500-25G 25500-500MG 25500-5G
76900-004EA 183.54 USD
76900-004 76900-006 76900-008 76900-010
2,6-Difluoro-4-(methylthio)benzoic acid ≥97% research grade
2,6-Difluoro-4-(methylthio)benzoic acid
Formula: C₈H₆F₂O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD23713122
CAS Number: 1428234-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR