2-Fluoro-3-methylbenzyl alcohol ≥97% research grade

Supplier: AOBChem USA

Warning

25419-25G 25419-5G
76899-966EA 502.87 USD
76899-966 76899-968
2-Fluoro-3-methylbenzyl alcohol ≥97% research grade
2-Fluoro-3-methylbenzyl alcohol
Formula: C₈H₉FO
MW: 140.155 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368780
CAS Number: 307975-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR